Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brix1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026396.3 | NP_080672.3 | 5 - 6 | 482 | 88 | |
GenBank mRNA | AK002985.1 | - | 5 - 6 | 369 | 88 | |
AK088313.1 | - | 5 - 6 | 466 | 88 | ||
AK168325.1 | - | 5 - 6 | 455 | 88 | ||
BC014832.1 | - | 5 - 6 | 452 | 4484024 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |