Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pop5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026398.4 | NP_080674.1 | 4 - 5 | 574 | 75 | |
GenBank mRNA | AK005290.1 | - | 3 - 4 | 330 | 75 | |
AK012523.1 | - | 332 | 75 | |||
AK013257.1 | - | 2 - 3 | 311 | 75 | ||
BC022670.1 | - | 4 - 5 | 598 | 4224973 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |