Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P411200 |
AK002363.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P411200 |
AK002363.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
478535 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
100176 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
669525 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
676582 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
791573 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
654164 |
AK077521.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
654164 |
AK145704.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
654164 |
BC019968.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
654164 |
BC024478.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
654164 |
NM_134139.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
847246 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
671832 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
67248 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
174116 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
44475 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P124176 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P160294 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P411200 |
AK002363.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P411200 |
AK002363.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P411200 |
AK002363.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P411200 |
AK002363.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917804 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917789 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917759 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917792 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917807 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917771 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917809 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917805 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917813 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917810 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917773 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917796 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917795 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917775 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917758 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917797 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917764 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917778 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917803 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917785 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917768 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917800 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917816 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917790 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917787 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917794 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917765 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917786 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917766 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917770 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917776 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917779 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917814 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917815 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917782 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26917762 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P124176 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06966462 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P160294 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P411200 |
AK002363.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P411200 |
AK002363.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P123621 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P160294 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P124176 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08267813 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08267805 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08267799 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08641890 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08267812 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917662 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917612 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917646 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917604 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917789 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917676 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917609 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917828 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917696 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917766 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917664 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917742 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917758 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917691 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917743 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917637 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917654 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917726 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917674 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917683 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917798 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917768 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917649 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917656 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917819 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917782 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917707 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917818 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917759 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917756 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917727 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917616 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917613 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917803 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917725 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917717 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917797 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917821 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917787 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917648 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917740 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917671 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917633 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917714 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917796 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917837 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917614 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917753 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917805 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917830 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917731 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917807 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917730 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917621 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917721 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917810 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917747 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917705 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917712 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917686 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917653 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917647 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917728 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917667 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917703 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917652 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917672 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917773 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917722 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917765 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917634 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917775 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917779 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917689 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917617 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917694 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917627 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917815 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917783 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917745 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917639 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917834 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917650 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917794 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917661 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917831 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917786 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917718 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917804 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917680 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917678 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917729 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917744 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917699 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917659 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917632 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917666 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917709 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917644 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917629 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917660 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917827 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917645 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917638 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917792 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917832 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917795 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917681 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917785 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917702 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917762 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917668 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917751 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917839 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917770 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917764 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917814 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917800 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917733 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26917738 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917747 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08532932 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917815 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08944495 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917795 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08538501 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917743 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917602 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917782 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917637 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917753 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917639 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917818 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917620 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917730 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917773 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917765 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917779 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917825 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917738 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917803 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917604 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917807 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917607 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917614 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917726 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917768 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917729 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917742 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917749 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917770 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917755 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08886529 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917634 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917748 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08626039 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917810 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917787 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917786 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08878504 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08285071 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917733 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917745 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917766 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08736625 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917756 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917823 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917629 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917636 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917775 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917789 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917612 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917796 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08886524 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917627 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917731 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917800 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917762 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08878502 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08974134 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08974141 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917624 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917797 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917814 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917616 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917805 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917821 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917744 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917804 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917609 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08886527 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917819 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08242585 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08973162 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917623 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08497739 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26917785 |
- |
- |