Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abtb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030251.3 | NP_084527.2 | 5 - 6 | 592 | 63 | |
GenBank mRNA | AB053477.1 | - | 5 - 6 | 523 | 63 | |
AK049100.1 | - | 5 - 6 | 508 | 63 | ||
AK088437.1 | - | 5 - 6 | 557 | 63 | ||
AK089474.1 | - | 3 - 4 | 956 | 63 | ||
AK220208.1 | - | 5 - 6 | 449 | 63 | ||
BC003234.1 | - | 5 - 6 | 536 | 3483893 | 63 | |
CO804154.1 | - | 4 - 5 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |