Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dqx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033606.3 | NP_291084.1 | 4 - 5 | 1184 | 74 | |
GenBank mRNA | AF318278.1 | - | 3 - 4 | 828 | 74 | |
BC075680.1 | - | 4 - 5 | 1184 | 30612145 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |