Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grid2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159321.1 | NP_001152793.1 | 19 - 20 | 3471 | 67 | |
NM_133355.1 | NP_579933.1 | 18 - 19 | 2775 | 67 | ||
GenBank mRNA | AF099933.2 | - | 18 - 19 | 2775 | 67 | |
BC075624.1 | - | 2827 | 6841572 | 67 | ||
DQ193535.1 | - | 19 - 20 | 3471 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |