Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mef2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133665.3 | NP_598426.1 | 8 - 9 | 1394 | 82 | |
GenBank mRNA | AK141270.1 | - | 8 - 9 | 1374 | 82 | |
AK155778.1 | - | 8 - 9 | 1395 | 82 | ||
BC011070.1 | - | 8 - 9 | 1362 | 3497896 | 82 | |
S68893.1 | - | 9 - 10 | 1237 | 82 | ||
S68895.1 | - | 8 - 9 | 1213 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |