Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20878432 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20878434 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20878431 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20878433 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P249051 |
AK167622.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P249051 |
AK187139.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P249051 |
BC006882.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P249051 |
AK150756.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P249051 |
AK152302.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P249051 |
AK167622.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P249051 |
AK187139.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P249051 |
BC006882.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P249051 |
AK150756.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P249051 |
AK152302.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
848470 |
AK017783.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
848470 |
AK084890.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
848470 |
AK087771.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
848470 |
NM_001081112.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
656145 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P249051 |
AK167622.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P249051 |
AK187139.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P249051 |
BC006882.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P249051 |
AK150756.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P249051 |
AK152302.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P249051 |
AK167622.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P249051 |
AK187139.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P249051 |
BC006882.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P249051 |
AK150756.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P249051 |
AK152302.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P249051 |
AK167622.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P249051 |
AK187139.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P249051 |
BC006882.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P249051 |
AK150756.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P249051 |
AK152302.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P249051 |
AK167622.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P249051 |
AK187139.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P249051 |
BC006882.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P249051 |
AK150756.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P249051 |
AK152302.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20878411 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20878439 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20878427 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20878431 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20878437 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20878413 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20878433 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20878434 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20878415 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20878407 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20878420 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20878421 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20878451 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20878449 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20878417 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20878419 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20878446 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20878441 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20878428 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20878424 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20878445 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20878443 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20878414 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20878453 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P249051 |
AK167622.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P249051 |
AK187139.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P249051 |
BC006882.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P249051 |
AK150756.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P249051 |
AK152302.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P249051 |
AK167622.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P249051 |
AK187139.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P249051 |
BC006882.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P249051 |
AK150756.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P249051 |
AK152302.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P133625 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P131717 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00700300 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00884632 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00884630 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20878433 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20878432 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20878434 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20878431 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878461 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878425 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878419 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878473 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878482 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878395 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878486 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878410 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878455 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878428 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878468 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878499 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878457 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878456 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878488 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878449 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878443 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878470 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878481 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878494 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878479 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878466 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878464 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878459 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878454 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878434 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878407 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878424 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878487 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878435 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878415 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878441 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878431 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878476 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878413 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878451 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878453 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878445 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878490 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878417 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878437 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878439 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878474 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878446 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878433 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878414 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20878429 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878455 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878419 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878453 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878411 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878481 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878428 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00791185 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878473 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878443 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01073203 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878445 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878422 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878439 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00100399 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878446 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878410 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878466 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878451 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878449 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878470 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878461 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878433 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00511267 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878415 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878413 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00700300 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00823470 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878471 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878416 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01065579 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00884632 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878407 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00961320 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878479 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00100425 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878420 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878437 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878474 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878475 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878457 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878421 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878414 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00433412 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20878425 |
- |
- |