Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2173968 |
AK142857.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2173968 |
AJ344340.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2173968 |
AK013115.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2173968 |
NM_133710.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P473953 |
AK171161.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P473953 |
AJ344340.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P473953 |
NM_133710.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P473953 |
AK171161.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P473953 |
AJ344340.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P473953 |
NM_133710.3 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
405064 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
408617 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
293632 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
198450 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
96853 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
124133 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
689182 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
725896 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
177839 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
698506 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
93867 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
194763 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
451312 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
81416 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
230036 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
510615 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
307967 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
565748 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
50260 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
317409 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
718669 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
889112 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
958761 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
954303 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
692013 |
AK029910.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
52445 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
712414 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
590548 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
606253 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
638859 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
6175 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
185604 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
856777 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
626132 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
852323 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
686608 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
336047 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
845545 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
940494 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
584914 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
486426 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
635278 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
798473 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
460293 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
417933 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
455163 |
AK049768.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
58452 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
145078 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
562704 |
AK036430.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
562704 |
AK157817.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
562704 |
BC040462.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
562704 |
BC051247.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
562704 |
BC066013.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
562704 |
NM_033523.4 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
210594 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
676602 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
171722 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
699961 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
492444 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
162109 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
385608 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
463966 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
789522 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
170101 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
558525 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
221233 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
638305 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
647968 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
815821 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
758210 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
3155 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
411092 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
657429 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
578933 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
391307 |
AB055389.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
391307 |
AK051184.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
391307 |
AK077219.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
391307 |
AK154131.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
391307 |
AK156412.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
391307 |
AK160129.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
391307 |
AK165956.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
391307 |
AK171699.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
391307 |
AK172120.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
391307 |
AK172432.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
391307 |
BC033460.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
391307 |
BC048377.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
391307 |
BC060503.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
391307 |
NM_080452.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
198075 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
114643 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
877548 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
161527 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
176618 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
66551 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
169042 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
19168 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
65629 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
139294 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
95426 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
50295 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
56263 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
52653 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
12123 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
108094 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P127763 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P160620 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P473953 |
AK171161.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P473953 |
AJ344340.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P473953 |
NM_133710.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P473953 |
AK171161.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P473953 |
AJ344340.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P473953 |
NM_133710.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2173968 |
AK142857.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2173968 |
AJ344340.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2173968 |
AK013115.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2173968 |
NM_133710.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P486943 |
AJ344340.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P486943 |
NM_133710.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P486943 |
AK138484.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P473953 |
AK171161.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P473953 |
AJ344340.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P473953 |
NM_133710.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P473953 |
AK171161.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P473953 |
AJ344340.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P473953 |
NM_133710.3 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P115126 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07643205 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07643193 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07643197 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08104345 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08104339 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08116200 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08149892 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07643201 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07643209 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07643208 |
- |
- |