Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nusap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042652.1 | NP_001036117.1 | 3 | 689 | 95 | |
NM_133851.3 | NP_598612.1 | 3 | 689 | 95 | ||
GenBank mRNA | AF305710.1 | - | 3 | 270 | 95 | |
AK042697.1 | - | 270 | 95 | |||
AK076028.1 | - | 3 | 259 | 95 | ||
AK088389.1 | - | 3 | 259 | 95 | ||
BC004787.1 | - | 3 | 239 | 3588697 | 95 | |
BC100392.1 | - | 3 | 689 | 30110191 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Amplicon spans exons and probe does not span exons |