Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080968.1 | NP_001074437.1 | 9 - 10 | 708 | 81 | |
NM_133852.2 | NP_598613.2 | 10 - 11 | 789 | 81 | ||
GenBank mRNA | AK076370.1 | - | 5 - 6 | 329 | 81 | |
AK077516.1 | - | 9 - 10 | 710 | 82 | ||
AK220509.1 | - | 10 - 11 | 717 | 81 | ||
BC011407.1 | - | 8 - 9 | 509 | 3596079 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |