Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133922.3 | NP_598683.3 | 11 - 12 | 1874 | 105 | |
GenBank mRNA | AB041543.1 | - | 9 - 10 | 1506 | 105 | |
AK029235.1 | - | 2 - 3 | 320 | 105 | ||
AK040525.1 | - | 11 - 12 | 1876 | 105 | ||
AK128931.1 | - | 11 - 12 | 1825 | 105 | ||
AK162934.1 | - | 11 - 12 | 1582 | 105 | ||
AK173281.1 | - | 11 - 12 | 3578 | 105 | ||
BC066816.1 | - | 11 - 12 | 1794 | 30084911 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |