Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133922.3 | NP_598683.3 | 16 - 17 | 2615 | 84 | |
GenBank mRNA | AB041543.1 | - | 14 - 15 | 2247 | 84 | |
AK029235.1 | - | 6 - 7 | 1613 | 84 | ||
AK038090.1 | - | 2 - 3 | 265 | 84 | ||
AK040525.1 | - | 16 - 17 | 2617 | 84 | ||
AK128931.1 | - | 16 - 17 | 2566 | 84 | ||
AK162934.1 | - | 16 - 17 | 85 | |||
AK173281.1 | - | 16 - 17 | 4319 | 84 | ||
BC066816.1 | - | 16 - 17 | 2535 | 30084911 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |