Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pot1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133931.4 | NP_598692.1 | 12 - 13 | 1565 | 123 | |
GenBank mRNA | AK036052.1 | - | 12 - 13 | 1566 | 123 | |
AK040864.1 | - | 12 - 13 | 1558 | 123 | ||
AK085041.1 | - | 12 - 13 | 1556 | 123 | ||
AK137376.1 | - | 12 - 13 | 1535 | 123 | ||
BC016121.1 | - | 12 - 13 | 1527 | 4501733 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |