Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133966.2 | NP_598727.1 | 3 - 4 | 378 | 58 | |
GenBank mRNA | AK033477.1 | - | 3 - 4 | 406 | 58 | |
AK089463.1 | - | 3 - 4 | 458 | 58 | ||
AK154832.1 | - | 3 - 4 | 458 | 58 | ||
BC013550.1 | - | 3 - 4 | 378 | 4222283 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |