Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk1g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159591.1 | NP_001153063.1 | 9 - 10 | 1321 | 110 | |
NM_134002.2 | NP_598763.1 | 8 - 9 | 1498 | 110 | ||
GenBank mRNA | AK077076.1 | - | 9 - 10 | 1301 | 110 | |
AK132871.1 | - | 9 - 10 | 1322 | 110 | ||
AK152413.1 | - | 8 - 9 | 1497 | 110 | ||
BC004839.1 | - | 8 - 9 | 1440 | 2648652 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |