Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100031053.mm.1 |
NM_134025.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100031053.mm.1 |
NM_134025.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100031053.mm.1 |
TC1100003391.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100031048.mm.1 |
NM_134025.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100031048.mm.1 |
NM_134025.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100031048.mm.1 |
TC1100003391.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100031052.mm.1 |
NM_134025.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100031052.mm.1 |
NM_134025.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100031052.mm.1 |
TC1100003391.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100031047.mm.1 |
NM_134025.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100031047.mm.1 |
NM_134025.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100031047.mm.1 |
TC1100003391.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100031061.mm.1 |
NM_134025.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100031061.mm.1 |
NM_134025.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100031061.mm.1 |
TC1100003391.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100031058.mm.1 |
NM_134025.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100031058.mm.1 |
NM_134025.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100031058.mm.1 |
TC1100003391.mm.1 |
MTA TC |