Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164743.1 | NP_001158215.1 | 9 - 10 | 1415 | 89 | |
NM_134069.3 | NP_598830.2 | 10 - 11 | 1539 | 89 | ||
GenBank mRNA | AK128896.1 | - | 10 - 11 | 1523 | 89 | |
AK143896.1 | - | 10 - 11 | 1511 | 89 | ||
BC018252.1 | - | 10 - 11 | 1468 | 4240254 | 89 | |
BC024459.1 | - | 10 - 11 | 1495 | 4975341 | 89 | |
BC034179.1 | - | 10 - 11 | 1359 | 4972500 | 89 | |
BC034183.1 | - | 10 - 11 | 1344 | 4973620 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |