Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134139.1 | NP_598900.1 | 2 - 3 | 171 | 91 | |
GenBank mRNA | AK077521.1 | - | 2 - 3 | 172 | 91 | |
AK145704.1 | - | 2 - 3 | 184 | 91 | ||
BC019968.1 | - | 2 - 3 | 171 | 4167260 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |