Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134139.1 | NP_598900.1 | 7 - 8 | 725 | 68 | |
GenBank mRNA | AK077521.1 | - | 7 - 8 | 726 | 68 | |
AK145704.1 | - | 7 - 8 | 606 | 68 | ||
BC019968.1 | - | 7 - 8 | 725 | 4167260 | 68 | |
BC024478.1 | - | 3 - 4 | 320 | 4977054 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |