Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cspg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139001.2 | NP_620570.2 | 1 - 2 | 176 | 96 | |
GenBank mRNA | AF352400.1 | - | 1 - 2 | 120 | 96 | |
AK052394.1 | - | 1 - 2 | 248 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Inventoried Probe spans exons |