Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alox12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009659.2 | NP_033789.1 | 5 - 6 | 829 | 67 | |
GenBank mRNA | AF059251.1 | - | 5 - 6 | 707 | 67 | |
BC113149.1 | - | 5 - 6 | 793 | 40054352 | 67 | |
Y14334.1 | - | 5 - 6 | 829 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |