Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023617.2 | NP_076106.2 | 18 - 19 | 2117 | 69 | |
GenBank mRNA | AF172276.2 | - | 18 - 19 | 2193 | 69 | |
AK004700.1 | - | 7 - 8 | 913 | 69 | ||
AK149434.1 | - | 18 - 19 | 2078 | 69 | ||
AY347569.1 | - | 18 - 19 | 1994 | 69 | ||
AY347570.1 | - | 18 - 19 | 1994 | 69 | ||
BC049853.1 | - | 18 - 19 | 2069 | 69 | ||
BC138876.1 | - | 18 - 19 | 2096 | 8861898 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |