Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025692.3 | NP_079968.2 | 8 - 9 | 986 | 113 | |
GenBank mRNA | AK017787.1 | - | 8 - 9 | 986 | 113 | |
AK077825.1 | - | 8 - 9 | 967 | 113 | ||
AK151823.1 | - | 8 - 9 | 943 | 113 | ||
AK152029.1 | - | 8 - 9 | 981 | 113 | ||
AK168907.1 | - | 8 - 9 | 980 | 113 | ||
BC019764.1 | - | 8 - 9 | 863 | 3989119 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |