Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Swt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025819.4 | NP_080095.3 | 8 - 9 | 1345 | 71 | |
GenBank mRNA | AK004803.2 | - | 8 - 9 | 1292 | 71 | |
BC055064.1 | - | 8 - 9 | 1346 | 6528686 | 71 | |
BC060204.1 | - | 8 - 9 | 1285 | 6855442 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |