Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtfr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253390.1 | NP_001240319.1 | 5 - 6 | 648 | 137 | |
NM_001253391.1 | NP_001240320.1 | 5 - 6 | 810 | 137 | ||
NM_026182.5 | NP_080458.1 | 5 - 6 | 658 | 137 | ||
NR_045575.1 | 6 - 7 | 695 | 137 | |||
GenBank mRNA | AF354708.1 | - | 4 - 5 | 609 | 137 | |
AK004846.1 | - | 5 - 6 | 636 | 137 | ||
AK006952.1 | - | 5 - 6 | 810 | 137 | ||
AK009195.2 | - | 5 - 6 | 624 | 137 | ||
AK012823.1 | - | 636 | 137 | |||
AK016316.1 | - | 5 - 6 | 798 | 137 | ||
AK018048.1 | - | 5 - 6 | 640 | 137 | ||
AK075822.1 | - | 5 - 6 | 634 | 137 | ||
AK075992.1 | - | 5 - 6 | 658 | 137 | ||
AK134822.1 | - | 6 - 7 | 652 | 137 | ||
AK161251.1 | - | 5 - 6 | 631 | 137 | ||
BC019971.1 | - | 5 - 6 | 606 | 4159093 | 137 | |
BC042471.1 | - | 4 - 5 | 582 | 3988195 | 137 | |
DQ369744.1 | - | 5 - 6 | 769 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |