Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ufl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026194.4 | NP_080470.2 | 7 - 8 | 731 | 108 | |
GenBank mRNA | AK007993.2 | - | 7 - 8 | 676 | 108 | |
AK032663.1 | - | 7 - 8 | 667 | 108 | ||
AK076206.1 | - | 7 - 8 | 676 | 108 | ||
AK129211.1 | - | 6 - 7 | 644 | 108 | ||
AK132697.1 | - | 7 - 8 | 695 | 108 | ||
BC080203.1 | - | 7 - 8 | 709 | 108 | ||
BC099529.1 | - | 7 - 8 | 719 | 108 | ||
BC132195.1 | - | 7 - 8 | 683 | 40130472 | 108 | |
BC138153.1 | - | 7 - 8 | 683 | 8861172 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |