Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Patl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026251.2 | NP_080527.2 | 4 - 5 | 493 | 75 | |
GenBank mRNA | AK015195.1 | - | 4 - 5 | 494 | 75 | |
BY714747.1 | - | 4 - 5 | 494 | 75 | ||
DV658440.1 | - | 2 - 3 | 104 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |