Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026283.2 | NP_080559.1 | 1 - 2 | 246 | 95 | |
GenBank mRNA | AK006211.1 | - | 1 - 2 | 247 | 95 | |
AK015593.1 | - | 1 - 2 | 225 | 95 | ||
AK075749.1 | - | 1 - 2 | 197 | 95 | ||
BC116745.1 | - | 1 - 2 | 234 | 40126064 | 95 | |
BC116747.1 | - | 1 - 2 | 234 | 40126066 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |