Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsto2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026619.2 | NP_080895.2 | 3 - 4 | 331 | 85 | |
NM_030051.1 | NP_084327.1 | 3 - 4 | 284 | 85 | ||
GenBank mRNA | AK018953.1 | - | 3 - 4 | 323 | 86 | |
AK019582.1 | - | 3 - 4 | 250 | 85 | ||
AK075753.1 | - | 3 - 4 | 332 | 85 | ||
AK077086.1 | - | 3 - 4 | 285 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Amplicon length between 86 and 100 Mus musculus Probe spans exons |