Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmbim6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171034.1 | NP_001164505.1 | 5 - 6 | 628 | 103 | |
NM_001171035.1 | NP_001164506.1 | 4 - 5 | 484 | 103 | ||
NM_001171036.1 | NP_001164507.1 | 5 - 6 | 449 | 103 | ||
NM_026669.4 | NP_080945.1 | 4 - 5 | 413 | 103 | ||
GenBank mRNA | AK019865.2 | - | 4 - 5 | 375 | 103 | |
AK049604.1 | - | 4 - 5 | 364 | 103 | ||
AK050299.1 | - | 4 - 5 | 376 | 103 | ||
AK050323.1 | - | 4 - 5 | 364 | 103 | ||
AK088030.1 | - | 4 - 5 | 375 | 103 | ||
AK088683.1 | - | 4 - 5 | 376 | 103 | ||
AK139901.1 | - | 4 - 5 | 376 | 103 | ||
AK159380.1 | - | 4 - 5 | 375 | 103 | ||
AK166935.1 | - | 4 - 5 | 375 | 103 | ||
BC005588.1 | - | 4 - 5 | 358 | 3708463 | 103 | |
BY753376.1 | - | 4 - 5 | 467 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |