Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pold4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027196.3 | NP_081472.1 | 1 - 2 | 294 | 86 | |
GenBank mRNA | AF515709.1 | - | 1 - 2 | 194 | 86 | |
AK010477.1 | - | 1 - 2 | 257 | 86 | ||
BC028520.1 | - | 1 - 2 | 215 | 1400386 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |