Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smg9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028047.2 | NP_082323.1 | 4 - 5 | 758 | 127 | |
GenBank mRNA | AK005126.1 | - | 4 - 5 | 758 | 127 | |
AK039368.1 | - | 4 - 5 | 757 | 127 | ||
AK141576.1 | - | 4 - 5 | 749 | 127 | ||
AK161454.1 | - | 4 - 5 | 768 | 127 | ||
AK169651.1 | - | 4 - 5 | 682 | 127 | ||
BC034211.1 | - | 4 - 5 | 673 | 4988361 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |