Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plxdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163608.1 | NP_001157080.1 | 6 - 7 | 868 | 69 | |
NM_028199.3 | NP_082475.3 | 6 - 7 | 847 | 69 | ||
GenBank mRNA | AF378760.1 | - | 6 - 7 | 781 | 69 | |
AK010361.1 | - | 6 - 7 | 847 | 69 | ||
AK036144.1 | - | 6 - 7 | 822 | 69 | ||
AK040620.1 | - | 5 - 6 | 724 | 69 | ||
AK153684.1 | - | 6 - 7 | 834 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |