Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbs5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028284.2 | NP_082560.1 | 10 - 11 | 931 | 66 | |
GenBank mRNA | AK006743.1 | - | 9 - 10 | 888 | 66 | |
AK012283.1 | - | 10 - 11 | 931 | 66 | ||
BC061031.1 | - | 10 - 11 | 932 | 6771365 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |