Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028303.3 | NP_082579.1 | 3 - 4 | 278 | 76 | |
GenBank mRNA | AK007454.1 | - | 3 - 4 | 233 | 76 | |
AK012624.1 | - | 3 - 4 | 279 | 76 | ||
BC004608.1 | - | 3 - 4 | 236 | 3585336 | 76 | |
BC022181.1 | - | 2 - 3 | 741 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |