Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028334.4 | NP_082610.1 | 1 - 2 | 97 | 81 | |
GenBank mRNA | AK012890.1 | - | 1 - 2 | 93 | 81 | |
CJ072834.1 | - | 1 - 2 | 98 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Inventoried Probe spans exons |