Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mon1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028369.3 | NP_082645.1 | 3 - 4 | 904 | 71 | |
GenBank mRNA | AK013387.1 | - | 3 - 4 | 851 | 71 | |
AK172624.1 | - | 3 - 4 | 811 | 71 | ||
BC048852.1 | - | 3 - 4 | 830 | 5148425 | 71 | |
BC058717.1 | - | 3 - 4 | 882 | 5704178 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |