Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028604.2 | NP_082880.2 | 2 - 3 | 231 | 105 | |
GenBank mRNA | AF532979.1 | - | 2 - 3 | 231 | 105 | |
AK010722.1 | - | 2 - 3 | 173 | 105 | ||
AK014181.1 | - | 2 - 3 | 161 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |