Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600009213.mm.1 |
NM_001079814.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009213.mm.1 |
NM_001079814.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009213.mm.1 |
NM_028720.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009213.mm.1 |
NM_028720.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009213.mm.1 |
TC1600001151.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600009211.mm.1 |
NM_001079814.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009211.mm.1 |
NM_001079814.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009211.mm.1 |
NM_028720.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009211.mm.1 |
NM_028720.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009211.mm.1 |
TC1600001151.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600009214.mm.1 |
NM_001079814.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009214.mm.1 |
NM_001079814.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009214.mm.1 |
NM_028720.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009214.mm.1 |
NM_028720.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009214.mm.1 |
TC1600001151.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600009215.mm.1 |
NM_001079814.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009215.mm.1 |
NM_001079814.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009215.mm.1 |
NM_028720.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009215.mm.1 |
NM_028720.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009215.mm.1 |
TC1600001151.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600009218.mm.1 |
NM_001079814.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009218.mm.1 |
NM_001079814.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009218.mm.1 |
NM_028720.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009218.mm.1 |
NM_028720.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009218.mm.1 |
TC1600001151.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600009219.mm.1 |
NM_001079814.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009219.mm.1 |
NM_001079814.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009219.mm.1 |
NM_028720.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009219.mm.1 |
NM_028720.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009219.mm.1 |
TC1600001151.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600009216.mm.1 |
NM_001079814.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009216.mm.1 |
NM_001079814.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009216.mm.1 |
NM_028720.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009216.mm.1 |
NM_028720.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009216.mm.1 |
TC1600001151.mm.1 |
MTA TC |