Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23269837 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23269838 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23269840 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23269839 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P297165 |
AK216219.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P297165 |
BC009141.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P297165 |
BC092058.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P297165 |
AK161646.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P297165 |
AK168966.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P297165 |
AK216219.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P297165 |
BC009141.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P297165 |
BC092058.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P297165 |
AK161646.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P297165 |
AK168966.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
214452 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
514392 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
314691 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
273981 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
964445 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
607348 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
600522 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
142754 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
325825 |
AK044619.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
325825 |
BC036966.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
325825 |
BC040465.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
325825 |
BC052329.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
325825 |
BC060083.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
325825 |
DQ176851.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
325825 |
NM_001113360.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
325825 |
NM_175556.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
844677 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
481820 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
48468 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
33907 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
17280 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P173957 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P297165 |
AK216219.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P297165 |
BC009141.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P297165 |
BC092058.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P297165 |
AK161646.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P297165 |
AK168966.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P297165 |
AK216219.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P297165 |
BC009141.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P297165 |
BC092058.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P297165 |
AK161646.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P297165 |
AK168966.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05387626 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P173957 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P297165 |
AK216219.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P297165 |
BC009141.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P297165 |
BC092058.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P297165 |
AK161646.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P297165 |
AK168966.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P297165 |
AK216219.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P297165 |
BC009141.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P297165 |
BC092058.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P297165 |
AK161646.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P297165 |
AK168966.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P108742 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P132208 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P100807 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P173957 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03318530 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03318525 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03318528 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03318526 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03318522 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23269838 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23269837 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23269840 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23269839 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269813 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269844 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269849 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23270029 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269815 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269820 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23270030 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23270017 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23270014 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269809 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269998 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269961 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23270024 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23270028 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269854 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269990 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23270010 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269950 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269810 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269821 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269948 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23270037 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269822 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269959 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269837 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269838 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269960 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269988 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269946 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269943 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269847 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269963 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269846 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269952 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23270007 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269973 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269993 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23270012 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269958 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269982 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23270033 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269808 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269968 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23270001 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269811 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269980 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269845 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269966 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269977 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269996 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23270005 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269954 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269953 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23270016 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23270022 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269957 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269976 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269835 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269818 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269940 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269944 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269806 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269965 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23270020 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23269999 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23270011 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23270032 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03340940 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23269854 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23269849 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03437977 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03437973 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03437979 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03340939 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23269968 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03335324 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23269852 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23269851 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03340942 |
- |
- |