Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg16l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205391.1 | NP_001192320.1 | 16 - 17 | 1818 | 59 | |
NM_001205392.1 | NP_001192321.1 | 14 - 15 | 1713 | 59 | ||
NM_029846.4 | NP_084122.2 | 15 - 16 | 1770 | 59 | ||
GenBank mRNA | AB087879.1 | - | 14 - 15 | 1524 | 59 | |
AB087880.1 | - | 15 - 16 | 1581 | 59 | ||
AB087881.1 | - | 16 - 17 | 1629 | 59 | ||
AB476646.1 | - | 16 - 17 | 1629 | 59 | ||
AK005181.1 | - | 15 - 16 | 1987 | 59 | ||
AK030983.1 | - | 15 - 16 | 1711 | 59 | ||
AK131120.1 | - | 13 - 14 | 2800 | 59 | ||
AK155358.1 | - | 14 - 15 | 1649 | 59 | ||
AK170073.1 | - | 14 - 15 | 1647 | 59 | ||
BC049122.1 | - | 16 - 17 | 1743 | 6813377 | 59 | |
BC052087.1 | - | 3 - 4 | 668 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |