Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps33a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029929.3 | NP_084205.3 | 11 - 12 | 1477 | 68 | |
GenBank mRNA | AF439858.1 | - | 11 - 12 | 1461 | 68 | |
AK019463.1 | - | 11 - 12 | 1461 | 68 | ||
AK041982.1 | - | 11 - 12 | 1473 | 68 | ||
AK140553.1 | - | 11 - 12 | 1466 | 68 | ||
AK150282.1 | - | 11 - 12 | 1469 | 68 | ||
AK150550.1 | - | 11 - 12 | 1469 | 68 | ||
BC046417.1 | - | 11 - 12 | 1468 | 4501207 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |