Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145824.4 | NP_665823.2 | 4 - 5 | 732 | 62 | |
GenBank mRNA | AK173177.1 | - | 4 - 5 | 545 | 62 | |
AY337314.1 | - | 4 - 5 | 733 | 62 | ||
BC024698.1 | - | 4 - 5 | 577 | 3667000 | 62 | |
EU281316.1 | - | 4 - 5 | 593 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |