Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007876.2 | NP_031902.2 | 7 - 8 | 1026 | 62 | |
GenBank mRNA | AK002380.1 | - | 6 - 7 | 907 | 62 | |
BC003492.1 | - | 7 - 8 | 896 | 2812088 | 62 | |
D13139.1 | - | 7 - 8 | 1026 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |