Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007883.2 | NP_031909.2 | 6 - 7 | 876 | 124 | |
GenBank mRNA | AB072269.1 | - | 6 - 7 | 904 | 124 | |
BC020144.1 | - | 6 - 7 | 770 | 124 | ||
BC044848.1 | - | 6 - 7 | 837 | 124 | ||
BC131653.1 | - | 6 - 7 | 852 | 40110529 | 124 | |
BC131654.1 | - | 6 - 7 | 852 | 40110531 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |