Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ecm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252653.1 | NP_001239582.1 | 2 - 3 | 255 | 126 | |
NM_007899.2 | NP_031925.2 | 2 - 3 | 255 | 126 | ||
GenBank mRNA | AK153009.1 | - | 2 - 3 | 254 | 126 | |
AK153471.1 | - | 2 - 3 | 254 | 126 | ||
AK159334.1 | - | 2 - 3 | 254 | 126 | ||
AK169939.1 | - | 2 - 3 | 257 | 126 | ||
BC138693.1 | - | 2 - 3 | 241 | 8861715 | 126 | |
L33416.1 | - | 2 - 3 | 248 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |