Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2020731 |
NM_009766.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2020731 |
AK038616.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
946163 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
463972 |
AJ243866.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
463972 |
BC010970.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
463972 |
BC040348.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
463972 |
NM_013921.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
63298 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
364104 |
AF027707.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
364104 |
AK156243.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
364104 |
AK168373.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
364104 |
AK171258.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
364104 |
BC030069.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
364104 |
NM_016778.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
157343 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P114453 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2020731 |
NM_009766.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2020731 |
AK038616.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P211416 |
BC106961.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P211416 |
BC106962.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P211416 |
NM_009766.3 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P211416 |
AK038616.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399201 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399209 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399234 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399205 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399162 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399168 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399204 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399158 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399210 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399197 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399159 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399189 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399177 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399186 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399187 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399173 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399184 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399182 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399192 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399174 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399195 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399235 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399185 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399202 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399170 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399191 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399193 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399237 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399166 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399161 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399176 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399167 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399203 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399207 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399198 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399155 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399171 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399165 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399160 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399200 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399180 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399232 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32399178 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09300648 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P114453 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P211416 |
BC106961.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P211416 |
BC106962.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P211416 |
NM_009766.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P211416 |
AK038616.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P137064 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P114453 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14739044 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P15005654 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14739045 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399209 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399254 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399207 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399235 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399148 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399282 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399242 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399186 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399161 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399269 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399202 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399182 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399244 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399293 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399165 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399247 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399154 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399160 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399212 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399240 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399177 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399297 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399283 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399281 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399272 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399152 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399255 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399268 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399214 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399232 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399230 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399176 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399184 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399279 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399249 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399299 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399215 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399145 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399198 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399180 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399292 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399286 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399228 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399248 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399221 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399275 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399265 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399142 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399284 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399201 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399140 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399200 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399195 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399223 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399251 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399294 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399131 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399259 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399245 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399170 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399143 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399267 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399138 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399276 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399167 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399226 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399136 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399155 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399173 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399205 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399204 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399192 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399169 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399134 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399189 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399133 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399264 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399126 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399216 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399178 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399288 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399241 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399158 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399159 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399151 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399229 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399278 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399257 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399237 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399193 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399218 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399213 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399168 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399262 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399135 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399127 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399187 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399253 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32399171 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15175160 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14840797 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399202 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399235 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399185 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399135 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399161 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14840791 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399201 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399173 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399154 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399140 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399176 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14840956 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399187 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399220 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399168 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399163 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14840396 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399136 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15175156 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399214 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399195 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399170 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399204 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399165 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399198 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399205 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399216 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399171 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14840770 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399193 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399229 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399183 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399192 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14840785 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399143 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399226 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399184 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399156 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399151 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399180 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15005654 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399207 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399224 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399223 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399182 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399174 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399209 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399215 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14840939 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399237 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399210 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399152 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399230 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14739037 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399221 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399200 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399142 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399218 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399160 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399134 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399178 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15199105 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32399138 |
- |
- |