Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043317.2 | NP_001036782.1 | 12 - 13 | 2549 | 65 | |
NM_009845.3 | NP_033975.3 | 10 - 11 | 2424 | 65 | ||
NR_102722.1 | 12 - 13 | 2621 | 65 | |||
NR_102723.1 | 12 - 13 | 2582 | 65 | |||
GenBank mRNA | AF102134.1 | - | 9 - 10 | 2193 | 65 | |
AF115400.1 | - | 9 - 10 | 2193 | 65 | ||
AF115401.1 | - | 9 - 10 | 2193 | 65 | ||
AK041230.1 | - | 12 - 13 | 2556 | 65 | ||
AK143835.1 | - | 12 - 13 | 2487 | 65 | ||
AK171589.1 | - | 9 - 10 | 2230 | 65 | ||
BC046957.1 | - | 12 - 13 | 2494 | 65 | ||
L16928.1 | - | 11 - 12 | 2672 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |