Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010046.2 | NP_034176.1 | 1 - 2 | 270 | 66 | |
GenBank mRNA | AB057816.1 | - | 1 - 2 | 258 | 66 | |
AF078752.1 | - | 1 - 2 | 328 | 66 | ||
BC003717.1 | - | 1 - 2 | 270 | 3486724 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |